Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat Prolactin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF1445SP
This item is not returnable.
View return policy
Description
Prolactin Polyclonal specifically detects Prolactin in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
Prolactin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
PRL, prolactin | |
Goat | |
Affinity Purified | |
RUO | |
5617 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Neutralization, ELISA | |
LYOPH | |
Western Blot 0.25 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.25-1.0 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
NP_035294 | |
PRL | |
E. coli-derived recombinant mouse Prolactin Leu32-Cys228 Accession # NP_035294 | |
25 μg | |
Primary | |
Detects mouse and rat Prolactin in Western blots. In sandwich immunoassays, less than 1% cross-reactivity with recombinant human Prolactin is observed. | |
Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction