Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat Prolactin Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB1445
This item is not returnable.
View return policy
Description
Prolactin Monoclonal specifically detects Prolactin in Mouse, Rat samples. It is validated for Western Blot.Specifications
Prolactin | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
CAA28018 | |
PRL | |
E. coli-derived recombinant mouse Prolactin Leu32-Cys228 Accession # CAA28018 | |
500 μg | |
Primary | |
Detects mouse and rat Prolactin in Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Prolactin is observed. | |
Mouse, Rat | |
Purified |
Western Blot | |
207518 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
PRL, prolactin | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5617 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction