Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat SCF/c-kit Ligand Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6998SP
This item is not returnable.
View return policy
Description
SCF/c-kit Ligand Polyclonal specifically detects SCF/c-kit Ligand in Mouse, Rat samples. It is validated for Western Blot.Specifications
SCF/c-kit Ligand | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor | |
Sheep | |
Affinity Purified | |
RUO | |
4254 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
P21581 | |
Kitl | |
E. coli-derived recombinant rat SCF/c-kit Ligand Gln26-Ala189 Accession # P21581 | |
25 μg | |
Primary | |
Detect mouse and rat SCF/c-kit Ligand in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human SCF/c-kit Ligand is observed. | |
Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction