Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat SMOC-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5550
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5550 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5550 Supplier R&D Systems Supplier No. AF5550
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

SMOC-1 Polyclonal specifically detects SMOC-1 in Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen SMOC-1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. AAD41590
Gene Alias Secreted modular calcium-binding protein 1, SMOC1, SMOC-1, SPARC related modular calcium binding 1, SPARC-related modular calcium-binding protein 1
Gene Symbols SMOC1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SMOC-1 His26-Val452 Accession # AAD41590
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64093
Test Specificity Detects mouse and rat SMOC-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human (rh) SMOC-1 is observed and less than 1% cross-reactivity with recombinant mouse SMOC-2 and rhSMOC-2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.