Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat SODD/BAG4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4349SP
This item is not returnable.
View return policy
Description
SODD/BAG4 Polyclonal specifically detects SODD/BAG4 in Mouse, Rat samples. It is validated for Western Blot.Specifications
SODD/BAG4 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BAG4, BAG-4, BAG-family molecular chaperone regulator-4, Bcl-2-associated athanogene 4, BCL2-associated athanogene 4, Silencer of death domains, SODDBAG family molecular chaperone regulator 4 | |
Goat | |
Antigen Affinity-purified | |
RUO | |
9530 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunoassay | |
LYOPH | |
Western Blot 1 ug/mL, Knockout Validated | |
Q8CI61 | |
BAG4 | |
E. coli-derived recombinant mouse SODD/BAG4 Met1-Leu457 Accession # Q8CI61 | |
25 μg | |
Primary | |
Detects endogenous mouse and rat SODD/BAG4 in Western blots. | |
Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction