Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat Tenascin R Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB1624SP
This item is not returnable.
View return policy
Description
Tenascin R Monoclonal specifically detects Tenascin R in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Tenascin R | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Janusin, MGC149328, Restrictin, tenascin R (restrictin, janusin), tenascin-R, TNR, TN-R | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7143 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Immunohistochemistry | |
619 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
TNR | |
ôrest L2ö fraction from bovine brain | |
25 μg | |
Primary | |
Detects mouse and rat Tenascin R in Western blots. | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction