Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat Testisin/Prss21 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6820
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6820 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6820 Supplier R&D Systems Supplier No. AF6820
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Testisin/Prss21 Polyclonal specifically detects Testisin/Prss21 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Testisin/Prss21
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q9JHJ7
Gene Alias EC 3.4.21, Eosinophil serine protease 1, ESP1EC 3.4.21.-, ESP-1serine protease from eosinophils, protease, serine, 21 (testisin), Prss21, Serine protease 21, TEST1, TEST1testisin, TESTISIN
Gene Symbols PRSS21
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Testisin/Prss21 Gln28-Asn298 Accession # Q9JHJ7
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10942
Test Specificity Detects recombinant mouse Testisin/Prss21 in direct ELISAs and Western blots. Detects mouse and rat Testisin/Prss21 in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) Prss8 and rmPrss27 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.