Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat Thioredoxin-like 5/TRP14 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3769
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3769 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3769 Supplier R&D Systems Supplier No. AF3769
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Thioredoxin-like 5/TRP14/TXNDC17 Polyclonal specifically detects Thioredoxin-like 5/TRP14/TXNDC17 in Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Thioredoxin-like 5/TRP14/TXNDC17
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Gene Accession No. Q9CQM5
Gene Alias MGC14353, Protein 42-9-9, thioredoxin (Trx)-related protein, 14 kDa, thioredoxin domain containing 17, thioredoxin domain-containing protein 17, Thioredoxin like 5, thioredoxin-like 5, Thioredoxin-like protein 5, TRP14, TRP1414 kDa thioredoxin-related protein, TXNDC17, TXNL5
Gene Symbols TXNDC17
Host Species Goat
Immunogen E. coli-derived recombinant mouse Thioredoxin-like 5/TRP14 Met1-Asp123 Accession # Q9CQM5
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84817
Test Specificity Detects endogenous mouse and rat Thioredoxin-like 5/TRP14 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.