Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat Thrombopoietin/Tpo Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF488NA
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF488NA 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF488NA Supplier R&D Systems Supplier No. AF488NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 4 publications

Thrombopoietin/THPO Polyclonal specifically detects Thrombopoietin/THPO in Mouse, Rat samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen Thrombopoietin/Tpo
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 0.1-0.3 ug/mL
Gene Alias Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF
Gene Symbols THPO
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Thrombopoietin/Tpo and S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Thrombopoietin/Tpo
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7066
Test Specificity Detects mouse and rat Thrombopoietin/Tpo in direct ELISAs. Neutralizes the biological activity of recombinant mouse Thrombopoietin/Tpo. It will also neutralize the activity of recombinant human (rh) Thrombopoietin/Tpo, although 25 times the amount of Ig is required. In direct ELISAs less than 15% cross-reactivity with rhTpo is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.