Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat Transglutaminase 2/TGM2 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF5418
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF5418 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF5418 Supplier R&D Systems Supplier No. BAF5418
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Transglutaminase 2/TGM2 Polyclonal specifically detects Transglutaminase 2/TGM2 in Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Transglutaminase 2/TGM2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot
Gene Accession No. P21981
Gene Alias C polypeptide, EC 2.3.2.13, G-ALPHA-h, protein-glutamine gamma-glutamyltransferase 2, protein-glutamine-gamma-glutamyltransferase, TG(C), TG2, TGase C, TGase H, TGase-2, TGase-H, TGC, TGCGNAH, TGM2, Tissue transglutaminase, Transglutaminase 2, transglutaminase 2 (C polypeptide, protein-glutamine-gamma-glutamyltransferase), Transglutaminase C, Transglutaminase H, Transglutaminase-2, tTG
Gene Symbols TGM2
Host Species Sheep
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Transglutaminase 2/TGM2 Ala2-Ala686 Accession # P21981
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7052
Test Specificity Detects mouse and rat Transglutaminase 2/TGM2 in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) TGM2 and rhTGM7 is observed, and less than 1% cross-reactivity with rhTGM3 and rhTGM4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.