Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat TRH-degrading Ectoenzyme/TRHDE Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2985SP
This item is not returnable.
View return policy
Description
TRH-degrading Ectoenzyme/TRHDE Polyclonal specifically detects TRH-degrading Ectoenzyme/TRHDE in Mouse, Rat samples. It is validated for Western Blot.Specifications
TRH-degrading Ectoenzyme/TRHDE | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.19.6, FLJ22381, PAP-II, PAP-IIthyroliberinase, PGPEP2, Pyroglutamyl-peptidase II, Thyroliberinase, thyrotropin-releasing hormone degrading enzyme, thyrotropin-releasing hormone-degrading ectoenzyme, TRHDE, TRH-DE, TRH-degrading ectoenzyme, TRHdegrading Ectoenzyme, TRH-DEthyrotropin-releasing hormone degrading ectoenzyme, TRH-specific aminopeptidase | |
Goat | |
Affinity Purified | |
RUO | |
29953 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q8K093 | |
TRHDE | |
Mouse myeloma cell line NS0-derived recombinant mouse TRH-degrading Ectoenzyme/TRHDE Arg64-His1025 Accession # Q8K093 | |
25 μg | |
Primary | |
Detects mouse and rat TRH-degrading Ectoenzyme/TRHDE in direct ELISAs and Western blots. | |
Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction