Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse/Rat TRH-degrading Ectoenzyme/TRHDE Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2985
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2985 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2985 Supplier R&D Systems Supplier No. AF2985
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

TRH-degrading Ectoenzyme/TRHDE Polyclonal specifically detects TRH-degrading Ectoenzyme/TRHDE in Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen TRH-degrading Ectoenzyme/TRHDE
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q8K093
Gene Alias EC 3.4.19.6, FLJ22381, PAP-II, PAP-IIthyroliberinase, PGPEP2, Pyroglutamyl-peptidase II, Thyroliberinase, thyrotropin-releasing hormone degrading enzyme, thyrotropin-releasing hormone-degrading ectoenzyme, TRHDE, TRH-DE, TRH-degrading ectoenzyme, TRHdegrading Ectoenzyme, TRH-DEthyrotropin-releasing hormone degrading ectoenzyme, TRH-specific aminopeptidase
Gene Symbols TRHDE
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TRH-degrading Ectoenzyme/TRHDE Arg64-His1025 Accession # Q8K093
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29953
Test Specificity Detects mouse and rat TRH-degrading Ectoenzyme/TRHDE in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.