Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat TSC22 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3779
This item is not returnable.
View return policy
Description
TSC22/TSC22D1 Polyclonal specifically detects TSC22/TSC22D1 in Mouse, Rat samples. It is validated for Western Blot.Specifications
TSC22/TSC22D1 | |
Polyclonal | |
Unconjugated | |
P62500.2 | |
TSC22D1 | |
E. coli-derived recombinant mouse TSC22 Met1-Ala143 Accession # P62500.2 | |
100 μg | |
Primary | |
Detects mouse and rat TSC22 in direct ELISAs and Western blots. | |
Mouse, Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Cerebral protein 2, Egr5, KIAA1994, MGC17597, Regulatory protein TSC-22, RP11-269C23.2, TGFB1I4, TGFB-stimulated clone 22 homolog, transforming growth factor beta 1 induced transcript 4, Transforming growth factor beta-1-induced transcript 4 protein, transforming growth factor beta-stimulated protein TSC-22, TSC22 domain family 1, TSC22 domain family protein 1, TSC22 domain family, member 1, TSC22D1, TSC22DKFZp686O19206 | |
Goat | |
Affinity Purified | |
RUO | |
8848 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction