Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat Wnt-5a Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 32 publications
Supplier: R&D Systems AF645
This item is not returnable.
View return policy
Description
Wnt-5a Polyclonal specifically detects Wnt-5a in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Wnt-5a | |
Polyclonal | |
Unconjugated | |
P22725 | |
WNT5A | |
E. coli-derived recombinant mouse Wnt-5a peptide Gln254-Cys334 Accession # P22725 | |
100 μg | |
Primary | |
Detects mouse and rat Wnt-5a in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse (rm) Wnt-5b is observed and less than 2% cross-reactivity with rmWnt-1, rmWnt-3a, rmWnt-4, rmWnt-8a, and rmWnt-10b is observed. | |
Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL | |
hWNT5A, protein Wnt-5a, wingless-type MMTV integration site family, member 5A, Wnt5a, WNT-5A protein | |
Goat | |
Affinity Purified | |
RUO | |
7474 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction