Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse RBP4/Retinol-Binding Protein 4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3476
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3476 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3476 Supplier R&D Systems Supplier No. AF3476
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

RBP4/Retinol-Binding Protein 4 Polyclonal specifically detects RBP4/Retinol-Binding Protein 4 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen RBP4/Retinol-Binding Protein 4
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL
Gene Accession No. Q00724
Gene Alias interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, RetinolBinding Protein 4, retinol-binding protein 4, plasma
Gene Symbols RBP4
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse RBP4 (R&D Systems, Catalog # 3476-LC) Glu19-Leu201 Accession # Q00724
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5950
Test Specificity Detects mouse RBP4 in direct ELISAs and Western blots. In these formats, approximately 10% cross-reactivity with recombinant human RBP4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.