Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Reelin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3820
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3820 100 μg
MAB3820SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3820 Supplier R&D Systems Supplier No. MAB3820
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Reelin Monoclonal specifically detects Reelin in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Reelin
Applications Western Blot
Classification Monoclonal
Clone 397919
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q60841
Gene Alias EC 3.4.21, EC 3.4.21.-, Reeler, reelin, RELN, RL, RLPRO1598
Gene Symbols RELN
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Reelin Ala27-Met862 Accession # Q60841
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5649
Test Specificity Detects mouse Reelin in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.