Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Reg1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1657
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1657 100 μg
AF1657SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1657 Supplier R&D Systems Supplier No. AF1657
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 2 publications

Reg1 Polyclonal specifically detects Reg1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Reg1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P43137
Gene Alias ICRF, Islet cells regeneration factor, Islet of Langerhans regenerating protein, lithostathine-1-alpha, Pancreatic stone protein, pancreatic stone protein, secretory, Pancreatic thread protein, protein-X, PSPregenerating islet-derived 1 alpha (pancreatic stone protein, pancreatic threadprotein), PSPS1REG-1-alpha, PSPSMGC12447, PTPP19, regenerating islet-derived 1 alpha, Regenerating islet-derived protein 1-alpha, Regenerating protein I alpha, REGlithostathine 1 alpha
Gene Symbols Reg1
Host Species Sheep
Immunogen E. coli-derived recombinant mouse Reg1 Gln22-Gly165 Accession # P43137
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5967
Test Specificity Detects mouse Reg1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse (rm) Reg2 is observed and less than 5% cross-reactivity with rmReg3A, recombinant human (rh) Reg1B, rhReg4, recombinant rat (rr) Reg3 and rrReg2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.