Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Reg2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB2098SP
This item is not returnable.
View return policy
Description
Reg2 Monoclonal specifically detects Reg2 in Mouse samples. It is validated for Western Blot.Specifications
Reg2 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
regenerating islet-derived 2, RegII | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5968 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
239237 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Reg2 | |
E. coli-derived recombinant mouse Reg2 | |
25 μg | |
Primary | |
Detects mouse Reg2 in direct ELISAs and Western blots. Does not cross-react with recombinant mouse (rm) Reg1, recombinant rat (rr) Reg2, rmReg3A, recombinant human Reg4, or rrReg4. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction