Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Reg3B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5110
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5110 Supplier R&D Systems Supplier No. MAB5110
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 6 publications

Reg3B Monoclonal specifically detects Reg3B in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Reg3B
Applications Western Blot
Classification Monoclonal
Clone 518630
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P35230
Gene Alias PAP1, Reg3B
Gene Symbols Reg3b
Host Species Rat
Immunogen E. coli-derived recombinant mouse Reg3B Glu27-Gly175 Accession # P35230
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18489
Test Specificity Detects mouse Reg3B in direct ELISAs and Western blots. In direct ELISAs, does not cross-react with recombinant mouse (rm) Reg3A, rmReg3D, recombinant human Reg3D, recombinant rat (rr) Reg3B, or rrReg3G.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.