Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Reg3B Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 6 publications
Supplier: R&D Systems MAB5110SP
This item is not returnable.
View return policy
Description
Reg3B Monoclonal specifically detects Reg3B in Mouse samples. It is validated for Western Blot.Specifications
Reg3B | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
P35230 | |
Reg3b | |
E. coli-derived recombinant mouse Reg3B Glu27-Gly175 Accession # P35230 | |
25 μg | |
Primary | |
Detects mouse Reg3B in direct ELISAs and Western blots. In direct ELISAs, does not cross-react with recombinant mouse (rm) Reg3A, rmReg3D, recombinant human Reg3D, recombinant rat (rr) Reg3B, or rrReg3G. | |
Mouse | |
Purified |
Western Blot | |
518630 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
PAP1, Reg3B | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
18489 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction