Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Reg3B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5110
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5110 Supplier R&D Systems Supplier No. AF5110
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 6 publications

Reg3B Polyclonal specifically detects Reg3B in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Reg3B
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. P35230
Gene Alias PAP1, Reg3B
Gene Symbols Reg3b
Host Species Sheep
Immunogen E. coli-derived recombinant mouse Reg3B Glu27-Gly175 Accession # P35230
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18489
Test Specificity Detects mouse Reg3B in direct ELISAs and Western blots. In direct ELISAs, approximately 75% cross-reactivity with recombinant rat (rr) Reg3B is observed. In Western blots approximately 30% cross-reactivity with rrReg3B is observed. In direct ELISAs and Western blots, less than 1% cross-reactivity with rrReg3A, recombinant mouse (rm) Reg3A, and rmReg3G is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.