Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Relaxin-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6637SP
This item is not returnable.
View return policy
Description
Relaxin-1 Polyclonal specifically detects Relaxin-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Relaxin-1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
bA12D24.3.1, bA12D24.3.2, H1, preprorelaxin H1, prorelaxin H1, relaxin 1, relaxin 1 (H1), relaxin H1, Relaxin1, RLN1, RLXH1 | |
Sheep | |
Affinity Purified | |
RUO | |
6013 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
NP_035402 | |
RLN1 | |
E. coli-derived recombinant mouse Relaxin-1 Arg23-Arg54 with an N-terminal Met (B chain) & Glu161-Cys185 (A chain) Accession # NP_035402 | |
25 μg | |
Primary | |
Detects mouse Relaxin-1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human Relaxin-1 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction