Learn More
Kent Scientific Mouse Retractor Set for SurgiSuite Multi-Functional Surgical Platforms
Shop All Kent Scientific Products
Description
Includes
Two 3cm tall magnetic fixators with two locking jaws, ten 10cm wire retractors, ten retractor tips (two 1mm sharp tip, two 1mm blunt tip, two 2.5mm blunt tip, two 5mm blunt tip, two 7.5mm blunt tip)
- Designed specifically for mouse
Specifications
Specifications
| For Use With (Application) | Surgery, Homeothermic Warming, Surgical Procedures, Tissue Retraction |
| Type | Retractor Set |
| For Use With (Equipment) | SurgiSuite and SurgiSuite-LG Multi-Functional Surgical Platforms |
| Includes | Two 3cm tall magnetic fixators with two locking jaws, ten 10cm wire retractors, ten retractor tips (two 1mm sharp tip, two 1mm blunt tip, two 2.5mm blunt tip, two 5mm blunt tip, two 7.5mm blunt tip) |