Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse RGM-C Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
$200.50 - $549.50
Specifications
Antigen | RGM-C/Hemojuvelin |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
RGM-C/Hemojuvelin Polyclonal specifically detects RGM-C/Hemojuvelin in Mouse samples. It is validated for Western Blot.Specifications
RGM-C/Hemojuvelin | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
DL-M, haemojuvelin, hemochromatosis type 2 (juvenile), hemojuvelin, HFE2, HFE2AMGC23953, HJV, HJVHemochromatosis type 2 protein, JH, repulsive guidance molecule c, RGMC, RGMCRGM domain family member C | |
HFE2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q7TQ32 | |
148738 | |
Mouse myeloma cell line NS0-derived recombinant mouse RGM-C isoform 1 (R&D Systems, Catalog # 3634-RG) Gln33-Asp393 (Ile379Val) Accession # Q7TQ32 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse RGM-C in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human RGM-C is observed and less than 5% cross-reactivity with recombinant mouse (rm) RGM-A and rmRGM-B is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title