Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse ROBO3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3155SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3155SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3155SP Supplier R&D Systems Supplier No. AF3155SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 6 publications

ROBO3 Polyclonal specifically detects ROBO3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ROBO3
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Z2I4
Gene Alias FLJ21044, HGPPS, HGPS, RBIG1horizontal gaze palsy with progressive scoliosis, retinoblastoma inhibiting gene 1, RIG1, roundabout (axon guidance receptor, Drosophila) homolog 3, roundabout homolog 3, roundabout, axon guidance receptor, homolog 3 (Drosophila), Roundabout-like protein 3
Gene Symbols ROBO3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived mouse ROBO3 Gly54-Ser545 Accession # Q9Z2I4
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64221
Test Specificity Detects mouse ROBO3 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) ROBO3 and less than 5% cross-reactivity with rhROBO2 and rhROBO4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.