Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse ROBO4 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $526.00
Specifications
Antigen | ROBO4 |
---|---|
Clone | 274914 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
MAB50041SP
![]() |
R&D Systems
MAB50041SP |
Unconjugated | 25 μg |
Each for $200.50
|
|
||||
MAB50041100
![]() |
R&D Systems
MAB50041100 |
Unconjugated | 100 μg |
Each for $526.00
|
|
||||
Description
ROBO4 Monoclonal specifically detects ROBO4 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
ROBO4 | |
Flow Cytometry 0.25 ug/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
ECSM4, FLJ20798, Magic roundabout, MGC133352, MGC133353, MRB, roundabout homolog 4, roundabout homolog 4, magic roundabout (Drosophila) | |
Y3 rat myeloid cell line transfected with mouse ROBO4 Leu28-Ser1012 Accession # Q8C310 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse ROBO4 in direct ELISAs. |
274914 | |
Flow Cytometry | |
Unconjugated | |
Rat | |
Mouse | |
Q8C310 | |
54538 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title