Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse S100A11 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5167SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5167SP 25 μg
MAB5167 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB5167SP Supplier R&D Systems Supplier No. MAB5167SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

S100A11 Monoclonal specifically detects S100A11 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen S100A11
Applications Western Blot
Classification Monoclonal
Clone 493804
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P50543
Gene Alias calgizzarin, Metastatic lymph node gene 70 protein, MLN 70, MLN70, protein S100-A11, Protein S100-C, S100 calcium binding protein A11, S100 calcium-binding protein A11, S100 calcium-binding protein A11 (calgizzarin), S100C, S100CS100 calcium binding protein A11 (calgizzarin)
Gene Symbols S100A11
Host Species Rat
Immunogen E. coli-derived recombinant mouse S100A11 Met1-Ile98 Accession # P50543
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6282
Test Specificity Detects mouse S100A11 in direct ELISAs and Western blots. In direct ELISAs, ;no cross-reactivity with recombinant mouse S100A1, 4, 6, 8, 9, 10, 13, or 16 is observed, and approximately 75% cross-reactivity with recombinant human (rh) S100A11 is observed. In Western blots, no cross-reactivity with rhS100A11 is observed
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.