Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse S100A8 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 17 publications
Supplier: R&D Systems AF3059SP
This item is not returnable.
View return policy
Description
S100A8 Polyclonal specifically detects S100A8 in Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
S100A8 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
60B8AG, CAGACP-10, calgranulin A, Calgranulin-A, Calprotectin L1L subunit, CFAG, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A | |
Goat | |
Affinity Purified | |
RUO | |
6279 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL, Immunocytochemistry 5-15 ug/mL | |
P27005 | |
S100A8 | |
E. coli-derived recombinant mouse S100A8 Pro2-Glu89 Accession # P27005 | |
25 μg | |
Primary | |
Detects mouse S100A8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse (rm) S100A9, rmS100A10, and recombinant human S100B is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction