Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse S1P1/EDG-1 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB7089A100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7089A100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7089A100 Supplier R&D Systems Supplier No. FAB7089A100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 5 publications

S1P1/EDG-1 Monoclonal specifically detects S1P1/EDG-1 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen S1P1/EDG-1
Applications Flow Cytometry
Classification Monoclonal
Clone 713412
Conjugate APC
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied in a saline solution containing BSA and sodium azide
Gene Accession No. O08530
Gene Alias CD363, CD363 antigen, CHEDG1, D1S3362, EDG1, EDG-1, EDG1edg-1, Endothelial differentiation G-protein coupled receptor 1, endothelial differentiation, sphingolipid G-protein-coupled receptor, 1, FLJ58121, S1P receptor 1, S1P receptor Edg-1, s1p1, S1P1ECGF1, S1PR1, sphingosine 1-phosphate receptor 1, sphingosine 1-phosphate receptor EDG1, Sphingosine 1-phosphate receptor Edg-1, sphingosine-1-phosphate receptor 1
Gene Symbols S1PR1
Host Species Rat
Immunogen Mouse S1P1/EDG-1 synthetic peptide (T4-H28), Accession No. O08530
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1901
Test Specificity Detects mouse S1P1/EDG-1 peptide in direct ELISAs.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.