Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse S1P1/EDG-1 Biotinylated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB7089B
This item is not returnable.
View return policy
Description
S1P1/EDG-1 Monoclonal specifically detects S1P1/EDG-1 in Mouse samples. It is validated for Flow Cytometry.Specifications
S1P1/EDG-1 | |
Monoclonal | |
Biotin | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD363, CD363 antigen, CHEDG1, D1S3362, EDG1, EDG-1, EDG1edg-1, Endothelial differentiation G-protein coupled receptor 1, endothelial differentiation, sphingolipid G-protein-coupled receptor, 1, FLJ58121, S1P receptor 1, S1P receptor Edg-1, s1p1, S1P1ECGF1, S1PR1, sphingosine 1-phosphate receptor 1, sphingosine 1-phosphate receptor EDG1, Sphingosine 1-phosphate receptor Edg-1, sphingosine-1-phosphate receptor 1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1901 | |
Mouse | |
Purified |
Flow Cytometry | |
713412 | |
Flow Cytometry 10 uL/10^6 cells | |
O08530 | |
S1PR1 | |
Mouse S1P 1EDG-1 synthetic peptide (T4-H28) Accession # O08530 | |
100 Tests | |
Primary | |
Detects mouse S1P 1EDG-1 peptide in direct ELISAs. | |
Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction