Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse S1P5/EDG-8 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB9084U100UG
This item is not returnable.
View return policy
Description
S1P5/EDG-8 Monoclonal specifically detects S1P5/EDG-8 in Mouse samples. It is validated for Flow Cytometry.Specifications
S1P5/EDG-8 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Edg-8, EDG8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, LPB4, S1P receptor 5, S1P5, S1PR5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2 | |
Mouse S1P 5EDG-8 peptide corresponding to the N-terminal extracellular sequence Accession # Q91X56 | |
RUO | |
53637 | |
Mouse | |
IgG |
Flow Cytometry | |
1196A | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q91X56 | |
Rabbit | |
100 μg | |
Primary | |
Detects mouse S1P 5EDG-8 in Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction