Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse S1P5/EDG-8 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $526.00
Specifications
Antigen | S1P5/EDG-8 |
---|---|
Clone | 1196A |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Description
S1P5/EDG-8 Monoclonal specifically detects S1P5/EDG-8 in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
S1P5/EDG-8 | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
Edg-8, EDG8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, LPB4, S1P receptor 5, S1P5, S1PR5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2 | |
S1pr5 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
1196A | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Mouse | |
Q91X56 | |
53637 | |
Mouse S1P 5EDG-8 peptide corresponding to the N-terminal extracellular sequence Accession # Q91X56 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse S1P 5EDG-8 in Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title