Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse S5a/Angiocidin Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5540
This item is not returnable.
View return policy
Description
S5a/Angiocidin Polyclonal specifically detects S5a/Angiocidin in Mouse samples. It is validated for Western Blot.Specifications
S5a/Angiocidin | |
Polyclonal | |
Unconjugated | |
O35226 | |
Goat | |
Affinity Purified | |
RUO | |
5710 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
PSMD4 | |
E. coli-derived recombinant mouse S5a/Angiocidin isoform A Val2-Glu254 Accession # O35226 | |
100 μg | |
Primary | |
Detects mouse S5a/Angiocidin in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction