Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Sca-1/Ly6 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Catalog No. FAB1226G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1226G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1226G Supplier R&D Systems Supplier No. FAB1226G
This item is not returnable. View return policy

Rat Monoclonal Antibody

Sca-1/Ly6 Monoclonal specifically detects Sca-1/Ly6 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Sca-1/Ly6
Applications Flow Cytometry
Classification Monoclonal
Clone 177228
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. CAA28351
Gene Alias C6orf23, G6D, Ly6, Ly-6, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25, Sca1
Gene Symbols Ly6a
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Sca-1/Ly6 C-terminally truncated Ly-6E allele Leu27-Gly119 Accession # CAA28351
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58530
Test Specificity Detects mouse Sca-1/Ly6 in direct ELISAs.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.