Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Sca-1/Ly6 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1226S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1226S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1226S100 Supplier R&D Systems Supplier No. FAB1226S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Sca-1/Ly6 Monoclonal specifically detects Sca-1/Ly6 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Sca-1/Ly6
Applications Flow Cytometry
Classification Monoclonal
Clone 177228
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. CAA28351
Gene Alias C6orf23, G6D, Ly6, Ly-6, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25, Sca1
Gene Symbols LY6A
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Sca-1/Ly6 C-terminally truncated Ly-6E allele Leu27-Gly119 Accession # CAA28351
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58530
Test Specificity Detects mouse Sca-1/Ly6 in direct ELISAs.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.