Learn More
Description
Specifications
Specifications
| Antigen | Sca-1/Ly6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 177228 |
| Conjugate | APC |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | CAA28351 |
| Gene Alias | C6orf23, G6D, Ly6, Ly-6, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25, Sca1 |
| Gene Symbols | Ly6a |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

