Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse SCARA5 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB4754
This item is not returnable.
View return policy
Description
SCARA5 Monoclonal specifically detects SCARA5 in Mouse samples. It is validated for Immunohistochemistry.Specifications
SCARA5 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
Q8K299 | |
SCARA5 | |
Mouse myeloma cell line NS0-derived recombinant mouse SCARA5 Arg83-Pro491 Accession # Q8K299 | |
100 μg | |
Primary | |
Detects mouse SCARA5 in direct ELISAs. In direct ELISAs, 100% cross-reactivty with recombinant human SCARA5 is observed. | |
Mouse | |
Purified |
Immunohistochemistry | |
462002 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ23907, MGC45780, scavenger receptor class A, member 5 (putative), Tesr, testis expressed scavenger receptor, UNQ2938/PRO28700 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
286133 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction