Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse SCARA5 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF4754
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF4754 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF4754 Supplier R&D Systems Supplier No. BAF4754
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

SCARA5 Polyclonal specifically detects SCARA5 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen SCARA5
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q8K299
Gene Alias FLJ23907, MGC45780, scavenger receptor class A, member 5 (putative), Tesr, testis expressed scavenger receptor, UNQ2938/PRO28700
Gene Symbols SCARA5
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse SCARA5 Arg83-Pro491 Accession # Q8K299
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286133
Test Specificity Detects mouse SCARA5 in Western blots. In Western blots, approximately 40% cross-reactivity with recombinant human SCARA5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.