Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse SCF, AOF Recombinant Protein

Catalog No. p-7203282
Encompass
Click to view available options
Quantity:
100 μg

E. coli Recombinant Protein

Stem cell factor (SCF) is the ligand of the c-Kit oncogene and is expressed by various structural and inflammatory cells in the airways. Binding of SCF by the c-Kit receptor leads to homodimerization of the receptor and the activation of signalling pathways such as PI-3, PLC-gamma, Jak/STAT, and MAP kinase pathways. SCF expression leads to the induction of mast cell survival and the expression and release of histamine, pro-inflammatory cytokines and chemokines. The inhibition of the SCF/c-Kit pathway leads to a decrease in histamine levels, mast cell and eosinophil infiltration, IL-4 production and airway hyperresponsiveness, suggesting this pathway may be a useful therapeutic target in inflammatory diseases such as asthma. At least two isoforms of SCF are known to exist.
  • Purity: >95% by SDS-Page
  • Endotoxin Concentration: AX41 ≤EUs/μg
  • Protein Length: 165 aa.
  • Biological Activity: <20 ng/mL
  • Molecular Weight: 18.4 kDa
  • Animal Origin Free: Yes
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P20826
For Use With (Application) Control, ELISA, Functional Assay, Gel Shift, Western Blot
Formulation 20 mM sodium phosphate with no preservative
Gene ID (Entrez) 17311
Molecular Weight (g/mol) 18.4 kDa
Name Mouse Stem Cell Factor (SCF), AOF
Quantity 100 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Gene Alias 710-712; blz; c-Kit ligand; Clo; cloud gray; Con; contrasted; CSF; DCUA; DFNA69; DKFZp686F2250; familial progressive hyperpigmentation 2; FPH2; FPHH; Gb; grizzle-belly; Hematopoietic growth factor KL; H-SCF; Kit ligand; Kitl; Kitlg; kit-ligand; KL-1; Mast cell growth factor; MGF; M-SCF; Processed kit ligand; SCF; SF; SHEP7; sKITLG; Sl; Slf; Soluble KIT ligand; Steel factor; steel factor/kit ligand; Stem cell factor; stem cell factor KL-1; stem cell factor, CSF
Gene Symbol Kitl
Specific Reactivity Mouse
Conjugate Unconjugated
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.