Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse SCF/c-kit Ligand Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AB455NA
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB455NA 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB455NA Supplier R&D Systems Supplier No. AB455NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

SCF/c-kit Ligand Polyclonal specifically detects SCF/c-kit Ligand in Mouse samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen SCF/c-kit Ligand
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 4-20 ug/mL
Gene Accession No. Q78ED8
Gene Alias c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor
Gene Symbols Kitl
Host Species Goat
Immunogen E. coli-derived recombinant mouse SCF/c-kit Ligand Lys26-Ala189 Accession # Q78ED8
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4254
Test Specificity Detects mouse SCF/c-kit Ligand in direct ELISAs and Western blots. In direct ELISAs, approximately 70% cross-reactivity with recombinant rat SCF/c-kit Ligand is observed, and approximately 25% cross-reactivity with recombinant human SCF/c-kit Ligand and recombinant feline SCF/c-kit Ligand is observed.
Reconstitution Reconstitute at 1 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.