Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse SCGF/CLEC11a Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$200.50 - $538.00
Specifications
Antigen | SCGF/CLEC11a |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
SCGF/CLEC11a Polyclonal specifically detects SCGF/CLEC11a in Mouse samples. It is validated for Western Blot.Specifications
SCGF/CLEC11a | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
CLEC11a, CLECSF3, LSLCL, P47, SCGF | |
Clec11a | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
O88200 | |
6320 | |
Mouse myeloma cell line NS0-derived recombinant mouse SCGF/CLEC11a Ala22-Phe328 Accession # O88200 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse SCGF/CLEC11a in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant human SCGF is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title