Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse SECTM1A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7837SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7837SP 25 μg
AF7837100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF7837SP Supplier R&D Systems Supplier No. AF7837SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Antibody

SECTM1A specifically detects SECTM1A in Mouse samples. It is validated for Immunohistochemistry, Blockade of Receptor-ligand Interaction.

Specifications

Antigen SECTM1A
Applications Immunohistochemistry, Blockade of Receptor-ligand Interaction
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction 0.2-1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q921W8
Gene Alias K12, SECTM
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SECTM1A Met1-Thr165 Accession # Q921W8
Purification Method Affinity Purified
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 6398
Test Specificity Detects mouse SECTM1A in direct ELISAs.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.