Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Semaphorin 3F Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3237SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3237SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3237SP Supplier R&D Systems Supplier No. MAB3237SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Semaphorin 3F Monoclonal specifically detects Semaphorin 3F in Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen Semaphorin 3F
Applications Immunohistochemistry
Classification Monoclonal
Clone 741307
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O88632
Gene Alias sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3F, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, 3F, sema III/F, sema IV, Sema3F, SEMA4, SEMA-IV, SEMAK, semaphorin III/F, semaphorin IV
Gene Symbols SEMA3F
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 3F Ala19 Pro775 (Arg583Ala and Arg586Ala) Accession # O88632
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6405
Test Specificity Detects mouse Semaphorin 3F in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Semaphorin 3A, 3B, 3C, 3E, 4C, 4D, 4G, recombinant human Semaphorin 3D or 3F is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.