Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Semaphorin 3F Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3237
This item is not returnable.
View return policy
Description
Semaphorin 3F Monoclonal specifically detects Semaphorin 3F in Mouse samples. It is validated for Immunohistochemistry.Specifications
Semaphorin 3F | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
O88632 | |
SEMA3F | |
Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 3F Ala19 Pro775 (Arg583Ala and Arg586Ala) Accession # O88632 | |
100 μg | |
Primary | |
Detects mouse Semaphorin 3F in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Semaphorin 3A, 3B, 3C, 3E, 4C, 4D, 4G, recombinant human Semaphorin 3D or 3F is observed. | |
Mouse | |
Purified |
Immunohistochemistry | |
741307 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3F, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, 3F, sema III/F, sema IV, Sema3F, SEMA4, SEMA-IV, SEMAK, semaphorin III/F, semaphorin IV | |
Rat | |
Protein A or G purified from ascites | |
RUO | |
6405 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction