Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Semaphorin 4C Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8497V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8497V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8497V100 Supplier R&D Systems Supplier No. FAB8497V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Semaphorin 4C Monoclonal specifically detects Semaphorin 4C in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Semaphorin 4C
Applications Flow Cytometry
Classification Monoclonal
Clone 663203
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q64151
Gene Alias FLJ20369, KIAA1739, MGC126382, MGC126383, M-Sema F, M-SEMA-F, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4C, SEMA4C, SEMACL1, SEMAF, SEMAI, semaphorin-4C
Gene Symbols SEMA4C
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 4C Ala21-Phe664 Accession # Q64151
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54910
Test Specificity Detects mouse Semaphorin 4C in direct ELISAs. Detects HEK293 human embryonic kidney cell line transfected with mouse Semaphorin 4C by Flow Cytometry. Does not detect untransfected or irrelevant transfected HEK293 cells.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.