Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse Semaphorin 4D/CD100 ELISA Kit

Catalog No. p-7219826
Encompass_Preferred
Click to view available options
Quantity:
5 x 96 Tests
96 Tests

Catalog No. EM68RBX5

Supplier: Invitrogen™ EM68RBX5

Only null left
Add to Cart
Add to Cart

ELISA

Mouse Semaphorin 4D/CD100 quantitates mouse Semaphorin 4D/CD100 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse Semaphorin 4D/CD100.

Semaphorins are neuronal chemorepellants that direct pioneering neurons during nervous system development. CD100 (also known as semaphorin 4d), a 150 kDa homodimer, is a novel semaphorin that induces B cells to aggregate and improves their viability in vitro. CD100 modifies CD40-CD40L B cell signaling by augmenting B cell aggregation and survival and down regulating CD23 expression. CD72 is a lymphocyte receptor for the class IV semaphorin CD100, which is involved in regulating B cell signaling. CD100 is expressed on resting and PHA stimulated T cells. The protein is weakly expressed on NK cells, EBV transformed B cells, monocytes and tumor/peripheral blood T cell lines, and at higher density on activated T cells and B cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number A8; BB18; C9orf164; CD100; coll-4; GR3; M-Sema G; M-sema-G; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4 D; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4 D; sema J; Sema4d; Semacl2; SEMAJ; semaphorin 4 D; semaphorin H; semaphorin-4 D; Semaphorin-C-like 2; semaphorin-J; Semcl2
Assay Range 23 to 5,000 pg/mL
Assay Sensitivity 23 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 20354
Gene Symbol Sema4d
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target CD100 (SEMA4D)
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.