Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Semaphorin 4F Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7200
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7200 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7200 Supplier R&D Systems Supplier No. MAB7200
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Semaphorin 4F Monoclonal specifically detects Semaphorin 4F in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.

Specifications

Antigen Semaphorin 4F
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 780225
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 uL/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready
Gene Accession No. Q9Z123
Gene Alias M-SEMA, m-Sema-M, PRO2353, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4F, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4F, Sema M, Sema W, SEMA4F, SEMAM, semaphorin-4F, semaphorin-M, Semaphorin-W, SEMAW, SEMAWsemaphorin M
Gene Symbols SEMA4F
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Sema-4F Arg41-Gly667 Accession # Q9Z123
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 10505
Test Specificity Detects mouse Sema-4F in ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.