Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Semaphorin 6C Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2108SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2108SP 25 μg
MAB2108 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2108SP Supplier R&D Systems Supplier No. MAB2108SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Semaphorin 6C Monoclonal specifically detects Semaphorin 6C in Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen Semaphorin 6C
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 256631
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9WTM3
Gene Alias KIAA1869, m-SemaY, m-SemaY2, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6C, Sema Y, Sema6C, Semaphorin Y, semaphorin-6C, semaphorin-Y, SEMAY
Gene Symbols SEMA6C
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 6C Ala26-Ile602 Accession # Q9WTM3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10500
Test Specificity Detects mouse Semaphorin 6C in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) Semaphorin 6C is observed and no cross-reactivity with rhSemaphorin 3A, 6B, recombinant mouse Semaphorin 3B, 3C, 3E, 3F, 6A, 6D, or 7A is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.