Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Semaphorin 6C Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2108
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2108 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2108 Supplier R&D Systems Supplier No. AF2108
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Semaphorin 6C Polyclonal specifically detects Semaphorin 6C in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Semaphorin 6C
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q9WTM3
Gene Alias KIAA1869, m-SemaY, m-SemaY2, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6C, Sema Y, Sema6C, Semaphorin Y, semaphorin-6C, semaphorin-Y, SEMAY
Gene Symbols SEMA6C
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 6C Ala26-Ile602 Accession # Q9WTM3
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10500
Test Specificity Detects mouse Semaphorin 6C in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human (rh) Semaphorin 6B and rhSemaphorin 6D is observed, 5% cross-reactivity with rhSemaphorin 6A is observed, and less than 3% cross-reactivity with rhSemaphorin 3A, rhSemaphorin 3B, recombinant mouse (rm) Semaphorin 3C, rmSemaphorin 3E, rmSemaphorin 3F, and rmSemaphorin 7A is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.