Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse SeP ELISA Kit (Colorimetric)
SDP

Catalog No. NB170625 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170625 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB170625 Supplier Novus Biologicals™ Supplier No. NBP318779
Add to Cart
Add to Cart
This item is not returnable. View return policy

Colorimetric ELISA Kit

Specifications

Assay Range 31.25-2000 pg/mL
Assay Sensitivity 18.75 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Biological Fluids, Plasma, Serum
Specificity This kit recognizes Mouse SeP in samples. No significant cross-reactivity or interference between Mouse SeP and analogues was observed.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 6414
Gene Symbol SELENOP
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym selenoprotein P, plasma, 1, SELP
Target Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.